The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106675.1 | NP_001100145.1 | 7 - 8 | 772 | 71 |
XM_006238507.1 | XP_006238569.1 | 5 - 6 | 703 | 71 |