Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rbfox1
Entrez Gene ID
302920
Gene Name
RNA binding protein, fox-1 homolog (C. elegans) 1
Gene Aliases
A2bp1
Chromosome Location
Chr.10: 8312961 - 8655426 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106974.2 NP_001100444.2 9 - 10 1170 111

View Full Product Details