Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Isoc2b
Entrez Gene ID
361501
Gene Name
isochorismatase domain containing 2b
Gene Aliases
Isoc2, RGD1309062
Chromosome Location
Chr.1: 72524503 - 72545331 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008367.1 NP_001008368.1 5 - 6 632 78
XM_006228274.2 XP_006228336.1 6 - 7 886 78
GenBank mRNA BC085336.1 - 5 - 6 632 78

View Full Product Details