Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cnppd1
Entrez Gene ID
316530
Gene Name
cyclin Pas1/PHO80 domain containing 1
Gene Aliases
RGD735175
Chromosome Location
Chr.9: 82338861 - 82345262 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199112.1 NP_954543.1 8 - 9 1010 81
XM_006245226.2 XP_006245288.1 8 - 9 1044 81
GenBank mRNA BC061750.1 - 8 - 9 1010 81

View Full Product Details