The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001030025.1 | NP_001025196.1 | 7 - 8 | 904 | 72 |
XM_006251483.2 | XP_006251545.1 | 8 - 9 | 1273 | 72 | |
XM_006251484.2 | XP_006251546.1 | 8 - 9 | 1179 | 72 | |
XM_006251485.2 | XP_006251547.1 | 8 - 9 | 1123 | 72 | |
XM_006251487.2 | XP_006251549.1 | 8 - 9 | 1270 | 72 | |
XM_008770270.1 | XP_008768492.1 | 8 - 9 | 1097 | 72 | |
GenBank mRNA | BC099752.1 | - | 7 - 8 | 904 | 72 |