Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zcchc24
Entrez Gene ID
361104
Gene Name
zinc finger, CCHC domain containing 24
Gene Aliases
RGD1306164
Chromosome Location
Chr.16: 1996540 - 2048292 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108394.1 NP_001101864.1 1 - 2 262 75

View Full Product Details