Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wnt5a
Entrez Gene ID
64566
Gene Name
wingless-type MMTV integration site family, member 5A
Gene Aliases
-
Chromosome Location
Chr.16: 4469451 - 4488707 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006252664.2 XP_006252726.1 2 - 3 239 94
XM_006252665.2 XP_006252727.1 2 - 3 201 94
XM_008771007.1 XP_008769229.1 2 - 3 1077 94
GenBank mRNA AB244721.1 - 1 - 2 135 94

View Full Product Details