Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sh2d4a
Entrez Gene ID
306376
Gene Name
SH2 domain containing 4A
Gene Aliases
-
Chromosome Location
Chr.16: 23084247 - 23156825 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012048.1 NP_001012048.1 2 - 3 408 65
XM_006253032.2 XP_006253094.1 2 - 3 421 65
GenBank mRNA BC079100.1 - 2 - 3 408 65

View Full Product Details