Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tfdp2
Entrez Gene ID
300947
Gene Name
transcription factor Dp-2 (E2F dimerization partner 2)
Gene Aliases
Tcfdp2
Chromosome Location
Chr.8: 104040796 - 104181228 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006243556.2 XP_006243618.1 - 450 161

View Full Product Details