The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009627.1 | NP_001009627.1 | 1 - 2 | 773 | 97 |
GenBank mRNA | BC088224.1 | - | 1 - 2 | 773 | 97 |