Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pkp2
Entrez Gene ID
287925
Gene Name
plakophilin 2
Gene Aliases
-
Chromosome Location
Chr.11: 88912163 - 88972213 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100499.1 NP_001093969.1 - 949 129

View Full Product Details