Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sdf2l1
Entrez Gene ID
680945
Gene Name
stromal cell-derived factor 2-like 1
Gene Aliases
-
Chromosome Location
Chr.11: 88122271 - 88124513 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109433.1 NP_001102903.1 2 - 3 435 57

View Full Product Details