The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199506.1 | NP_955800.1 | 1 - 1 | 1912 | 63 |
GenBank mRNA | AB113389.1 | - | 1 - 1 | 1912 | 63 |
AY573980.1 | - | 1 - 1 | 1861 | 63 |