The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006241090.1 | XP_006241152.1 | 5 - 6 | 627 | 82 |
XM_006241091.2 | XP_006241153.1 | 5 - 6 | 736 | 82 | |
XM_006241092.1 | XP_006241154.1 | 4 - 5 | 539 | 82 | |
XM_008765183.1 | XP_008763405.1 | 4 - 5 | 967 | 82 | |
XM_234837.7 | XP_234837.3 | 5 - 6 | 1068 | 82 | |
GenBank mRNA | FQ210490.1 | - | 5 - 6 | 632 | 82 |