Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmsbl1
Entrez Gene ID
286978
Gene Name
thymosin beta-like protein 1
Gene Aliases
-
Chromosome Location
Chr.X: 107612518 - 107614632 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173313.1 NP_775435.1 2 - 3 200 185
GenBank mRNA U25684.1 - 2 - 3 200 185

View Full Product Details