Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ankrd2
Entrez Gene ID
309374
Gene Name
ankyrin repeat domain 2 (stretch responsive muscle)
Gene Aliases
-
Chromosome Location
Chr.1: 261281859 - 261291307 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107589.1 NP_001101059.1 2 - 3 189 123
GenBank mRNA FQ216123.1 - 2 - 3 199 123
FQ216298.1 - 2 - 3 199 123
FQ216609.1 - 2 - 3 201 123
FQ216805.1 - 2 - 3 199 123

View Full Product Details