Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Usf1
Entrez Gene ID
83586
Gene Name
upstream transcription factor 1
Gene Aliases
-
Chromosome Location
Chr.13: 89797750 - 89805561 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031777.2 NP_113965.1 6 - 7 607 89
XM_006250266.2 XP_006250328.1 6 - 7 792 89
XM_008769765.1 XP_008767987.1 4 - 5 443 89
GenBank mRNA AF026476.1 - 4 - 5 476 89
BC088849.1 - 6 - 7 607 89

View Full Product Details