Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Casq1
Entrez Gene ID
686019
Gene Name
calsequestrin 1 (fast-twitch, skeletal muscle)
Gene Aliases
-
Chromosome Location
Chr.13: 90592676 - 90602365 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159594.1 NP_001153066.1 6 - 7 948 102
GenBank mRNA FM056035.1 - 3 - 4 253 102
FQ216689.1 - 6 - 7 525 102
FQ224875.1 - 7 - 8 775 102

View Full Product Details