Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc19
Entrez Gene ID
304984
Gene Name
coiled-coil domain containing 19
Gene Aliases
-
Chromosome Location
Chr.13: 90909480 - 90932958 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024882.1 NP_001020053.1 2 - 3 165 70
XM_006250296.2 XP_006250358.1 2 - 3 260 70
GenBank mRNA BC098058.1 - 2 - 3 165 70

View Full Product Details