The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080910.3 | NP_543186.1 | 3 - 4 | 309 | 76 |
XM_006250852.2 | XP_006250914.1 | 3 - 4 | 357 | 76 | |
XM_006250853.2 | XP_006250915.1 | 3 - 4 | 328 | 76 | |
XM_006250854.2 | XP_006250916.1 | 3 - 4 | 358 | 76 | |
GenBank mRNA | BC072508.1 | - | 3 - 4 | 299 | 76 |
BC085711.1 | - | 3 - 4 | 309 | 76 | |
D37979.1 | - | 1 - 2 | 214 | 76 |