Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxw4
Entrez Gene ID
309444
Gene Name
F-box and WD repeat domain containing 4
Gene Aliases
-
Chromosome Location
Chr.1: 265318526 - 265420503 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107600.1 NP_001101070.1 4 - 5 822 97

View Full Product Details