Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Shroom3
Entrez Gene ID
305230
Gene Name
shroom family member 3
Gene Aliases
RGD1310470, Shrm
Chromosome Location
Chr.14: 16622767 - 16903278 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100889.1 NP_001094359.1 5 - 6 3786 116
XM_006250723.2 XP_006250785.1 5 - 6 4797 116
XM_006250725.2 XP_006250787.1 3 - 4 6570 116

View Full Product Details