Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam47e
Entrez Gene ID
686140
Gene Name
family with sequence similarity 47, member E
Gene Aliases
Fam47e-ps1
Chromosome Location
Chr.14: 16991496 - 17027568 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008770096.1 XP_008768318.1 2 - 3 585 73

View Full Product Details