The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105976.1 | NP_001099446.1 | 3 - 4 | 463 | 89 |
GenBank mRNA | FQ217742.1 | - | 3 - 4 | 270 | 89 |
FQ229252.1 | - | 3 - 4 | 225 | 89 |