Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Arhgap30
Entrez Gene ID
498282
Gene Name
Rho GTPase activating protein 30
Gene Aliases
RGD1561419
Chromosome Location
Chr.13: 89774764 - 89796113 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109077.1 NP_001102547.1 11 - 12 1796 72
GenBank mRNA FQ211641.1 - - 1804 72

View Full Product Details