Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Samd10
Entrez Gene ID
499957
Gene Name
sterile alpha motif domain containing 10
Gene Aliases
RGD1563203
Chromosome Location
Chr.3: 177091467 - 177095598 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191967.1 NP_001178896.1 4 - 5 587 154
XM_006235798.1 XP_006235860.1 4 - 5 598 154

View Full Product Details