Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Echdc2
Entrez Gene ID
298381
Gene Name
enoyl CoA hydratase domain containing 2
Gene Aliases
RGD1308525
Chromosome Location
Chr.5: 127770570 - 127789568 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106675.1 NP_001100145.1 4 - 5 436 148

View Full Product Details