Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zw10
Entrez Gene ID
363059
Gene Name
zw10 kinetochore protein
Gene Aliases
-
Chromosome Location
Chr.8: 53365881 - 53390742 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024801.1 NP_001019972.1 2 - 3 325 107
GenBank mRNA BC097452.1 - 2 - 3 325 107

View Full Product Details