Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Peak1
Entrez Gene ID
315686
Gene Name
pseudopodium-enriched atypical kinase 1
Gene Aliases
RGD1312026
Chromosome Location
Chr.8: 60861742 - 61080046 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108149.1 NP_001101619.1 6 - 7 4505 68
XM_006243115.2 XP_006243177.1 5 - 6 4375 68
XM_006243116.2 XP_006243178.1 7 - 8 5127 68

View Full Product Details