Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Yes1
Entrez Gene ID
24884
Gene Name
Yamaguchi sarcoma viral (v-yes) oncogene homolog 1
Gene Aliases
Yes, c-Yes, p60c-yes, p61-Yes
Chromosome Location
Chr.9: 121802640 - 121877879 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033298.1 NP_150640.1 6 - 7 948 91
XM_006245675.2 XP_006245737.1 6 - 7 961 91
GenBank mRNA AB037472.1 - 5 - 6 725 91
BC079403.1 - 6 - 7 948 91

View Full Product Details