Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dgat2l6
Entrez Gene ID
678749
Gene Name
diacylglycerol O-acyltransferase 2-like 6
Gene Aliases
-
Chromosome Location
Chr.X: 70354357 - 70377887 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109370.1 NP_001102840.1 6 - 7 959 165

View Full Product Details