Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Haus8
Entrez Gene ID
290626
Gene Name
HAUS augmin-like complex, subunit 8
Gene Aliases
Ny-sar-48, RGD1311565
Chromosome Location
Chr.16: 19654785 - 19669213 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024971.2 NP_001020142.2 5 - 6 485 82
XM_008771074.1 XP_008769296.1 6 - 7 627 82
XM_008771075.1 XP_008769297.1 6 - 7 624 82
XM_008771076.1 XP_008769298.1 6 - 7 624 82
XM_008771077.1 XP_008769299.1 5 - 6 530 82
XM_008771078.1 XP_008769300.1 2 - 3 148 82
GenBank mRNA BC091198.1 - 5 - 6 474 82
DY317896.1 - 5 - 6 445 82
FQ225627.1 - 5 - 6 497 82

View Full Product Details