Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Haus8
Entrez Gene ID
290626
Gene Name
HAUS augmin-like complex, subunit 8
Gene Aliases
Ny-sar-48, RGD1311565
Chromosome Location
Chr.16: 19654785 - 19669213 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024971.2 NP_001020142.2 9 - 10 963 120
XM_008771074.1 XP_008769296.1 10 - 11 1105 120
XM_008771075.1 XP_008769297.1 10 - 11 1102 120
XM_008771076.1 XP_008769298.1 10 - 11 1102 120
XM_008771077.1 XP_008769299.1 9 - 10 1008 120
XM_008771078.1 XP_008769300.1 6 - 7 626 120
GenBank mRNA BC091198.1 - 9 - 10 950 120
BQ207735.1 - 2 - 3 120
FQ225627.1 - 9 - 10 977 120

View Full Product Details