Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ghitm
Entrez Gene ID
290596
Gene Name
growth hormone inducible transmembrane protein
Gene Aliases
MICS1
Chromosome Location
Chr.16: 14370269 - 14382647 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005908.1 NP_001005908.1 3 - 4 367 84
XM_006252786.2 XP_006252848.1 3 - 4 621 84
GenBank mRNA BC083778.1 - 3 - 4 367 84
FQ212260.1 - 3 - 4 567 84
FQ217853.1 - 3 - 4 386 84

View Full Product Details