The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005908.1 | NP_001005908.1 | 8 - 9 | 1094 | 93 |
XM_006252786.2 | XP_006252848.1 | 8 - 9 | 1348 | 93 | |
GenBank mRNA | BC083778.1 | - | 8 - 9 | 1094 | 93 |
FQ212260.1 | - | 8 - 9 | 1294 | 93 | |
FQ217853.1 | - | 8 - 9 | 1112 | 93 |