Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Babam1
Entrez Gene ID
290631
Gene Name
BRISC and BRCA1 A complex member 1
Gene Aliases
Merit40
Chromosome Location
Chr.16: 19792822 - 19798893 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006964.1 NP_001006965.1 7 - 8 895 115
XM_006252857.2 XP_006252919.1 7 - 8 919 115
GenBank mRNA BC083685.1 - 7 - 8 895 115
FQ214886.1 - 7 - 8 886 115
FQ233705.1 - 7 - 8 906 115

View Full Product Details