Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tatdn2
Entrez Gene ID
500295
Gene Name
TatD DNase domain containing 2
Gene Aliases
-
Chromosome Location
Chr.4: 145653300 - 145669343 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109252.1 NP_001102722.1 6 - 7 2365 113

View Full Product Details