Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gramd2
Entrez Gene ID
300761
Gene Name
GRAM domain containing 2
Gene Aliases
RGD1564007
Chromosome Location
Chr.8: 64503021 - 64539149 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006243197.2 XP_006243259.1 4 - 5 565 94
XM_006243198.2 XP_006243260.1 5 - 6 926 94
XM_006243199.2 XP_006243261.1 4 - 5 974 94
XM_006243200.2 XP_006243262.1 4 - 5 1314 94
XM_006243201.2 XP_006243263.1 4 - 5 2775 94
XM_008766374.1 XP_008764596.1 4 - 5 445 94
XM_217153.7 XP_217153.5 4 - 5 364 94

View Full Product Details