Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gramd2
Entrez Gene ID
300761
Gene Name
GRAM domain containing 2
Gene Aliases
RGD1564007
Chromosome Location
Chr.8: 64503021 - 64539149 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006243197.2 XP_006243259.1 8 - 9 905 96
XM_006243198.2 XP_006243260.1 9 - 10 1266 96
XM_006243199.2 XP_006243261.1 8 - 9 1314 96
XM_006243200.2 XP_006243262.1 8 - 9 1654 96
XM_006243201.2 XP_006243263.1 8 - 9 3115 96
XM_008766374.1 XP_008764596.1 8 - 9 785 96
XM_217153.7 XP_217153.5 8 - 9 704 96

View Full Product Details