Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gdap1l1
Entrez Gene ID
311616
Gene Name
ganglioside-induced differentiation-associated protein 1-like 1
Gene Aliases
Gdap1l
Chromosome Location
Chr.3: 159823755 - 159843238 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107798.2 NP_001101268.1 2 - 3 388 81
GenBank mRNA BC168159.1 - 2 - 3 388 81
FQ212886.1 - 2 - 3 445 81

View Full Product Details