Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prkx
Entrez Gene ID
501563
Gene Name
protein kinase, X-linked
Gene Aliases
-
Chromosome Location
Chr.X: 45080963 - 45124270 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033963.1 NP_001029135.1 8 - 9 1356 147
GenBank mRNA BC091203.1 - 8 - 9 1356 147

View Full Product Details