Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pmm2
Entrez Gene ID
302915
Gene Name
phosphomannomutase 2
Gene Aliases
-
Chromosome Location
Chr.10: 7056258 - 7077443 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106973.1 NP_001100443.1 6 - 7 573 62
XM_006245755.1 XP_006245817.1 7 - 8 912 62
GenBank mRNA BC168922.1 - 6 - 7 529 62
FQ232490.1 - 1 - 2 67 62

View Full Product Details