Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atp2b2
Entrez Gene ID
24215
Gene Name
ATPase, Ca++ transporting, plasma membrane 2
Gene Aliases
PMCA2
Chromosome Location
Chr.4: 145704779 - 145948997 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012508.5 NP_036640.1 - 2065 58
GenBank mRNA J03754.1 - - 2071 58

View Full Product Details