The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053914.1 | NP_446366.1 | 3 - 4 | 781 | 83 |
GenBank mRNA | AM419015.1 | - | 3 - 4 | 781 | 83 |
U90215.1 | - | 3 - 4 | 507 | 83 |