Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem26
Entrez Gene ID
309724
Gene Name
transmembrane protein 26
Gene Aliases
-
Chromosome Location
Chr.20: 21640655 - 21689655 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107623.1 NP_001101093.1 4 - 5 851 94

View Full Product Details