The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_147138.2 | NP_671479.1 | 3 - 4 | 342 | 78 |
GenBank mRNA | AF439397.1 | - | 3 - 4 | 324 | 78 |
AF532776.1 | - | 3 - 4 | 342 | 78 | |
BC063144.1 | - | 3 - 4 | 334 | 78 | |
EU000468.1 | - | 3 - 4 | 310 | 78 | |
FQ228814.1 | - | 1 - 2 | 76 | 78 |