Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cfp
Entrez Gene ID
299314
Gene Name
complement factor properdin
Gene Aliases
Pfc
Chromosome Location
Chr.X: 1311121 - 1316683 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106757.2 NP_001100227.1 2 - 3 235 110
GenBank mRNA BC158889.1 - 2 - 3 235 110
FQ209917.1 - 2 - 3 266 110
FQ212442.1 - 2 - 3 270 110

View Full Product Details