Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rgn
Entrez Gene ID
25106
Gene Name
regucalcin (senescence marker protein-30)
Gene Aliases
GNL, Rc, Reguc
Chromosome Location
Chr.X: 1833484 - 1848904 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031546.1 NP_113734.1 4 - 5 664 127
GenBank mRNA AB037934.1 - 4 - 5 664 127
BC078794.1 - 4 - 5 659 127
D38467.1 - 4 - 5 127
FQ219778.1 - 4 - 5 667 127
X69021.1 - 4 - 5 653 127

View Full Product Details