Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfhx3
Entrez Gene ID
307829
Gene Name
zinc finger homeobox 3
Gene Aliases
Atbf1, RGD1560268
Chromosome Location
Chr.19: 42753983 - 42925275 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008772567.1 XP_008770789.1 4 - 5 8489 101

View Full Product Details