Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gins2
Entrez Gene ID
292058
Gene Name
GINS complex subunit 2 (Psf2 homolog)
Gene Aliases
RGD1311055
Chromosome Location
Chr.19: 54151048 - 54163697 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106190.1 NP_001099660.1 3 - 4 588 71
XM_006255735.2 XP_006255797.1 3 - 4 629 71

View Full Product Details